(2R,3R)-1-Cbz-2-methylpiperidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR471275-1G OR471275-250MG
APOSOR471275-1GEA 1420 EUR
APOSOR471275-1G APOSOR471275-250MG
(2R,3R)-1-Cbz-2-methylpiperidine-3-carboxylic acid
(2R,3R)-1-Cbz-2-methylpiperidine-3-carboxylic acid
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277.316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28505589
CAS-Nummer: 1932256-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR