2-(4-Chlorophenyl)oxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471284-1G OR471284-250MG OR471284-5G
APOSOR471284-1GEA 601 EUR
APOSOR471284-1G APOSOR471284-250MG APOSOR471284-5G
2-(4-Chlorophenyl)oxazole-4-carboxylic acid
2-(4-Chlorophenyl)oxazole-4-carboxylic acid
Formel: C₁₀H₆ClNO₃
Molecular Weight: 223.613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16698588
CAS-Nummer: 1065102-51-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR