7-Chloro-2H-chromen-2-one

Lieferant: Apollo Scientific

OR471286-1G OR471286-250MG
APOSOR471286-1GEA 804 EUR
APOSOR471286-1G APOSOR471286-250MG
7-Chloro-2H-chromen-2-one
7-Chloro-2H-chromen-2-one
Formel: C₉H₅ClO₂
Molecular Weight: 180,588 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00152031
CAS-Nummer: 19063-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR