6-(Aminomethyl)-2-methylbenzoxazole

Lieferant: Apollo Scientific
Achtung

OR471294-1G OR471294-250MG
APOSOR471294-1GEA 2530 EUR
APOSOR471294-1G APOSOR471294-250MG
6-(Aminomethyl)-2-methylbenzoxazole
6-(Aminomethyl)-2-methylbenzoxazole
Formel: C₉H₁₀N₂O
Molecular Weight: 162,189 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26517728
CAS-Nummer: 1368311-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR