D-tert-Leucine methyl ester hydrochloride
Lieferant: Apollo Scientific
Synonyme:
H-D-3-Me-Val-OMe·HCl, (R)-Methyl 2-amino-3,3-dimethylbutanoate hydrochloride
OR471306-1G
OR471306-250MG
OR471306-5G
APOSOR471306-1GEA
92.8
EUR
APOSOR471306-1G
APOSOR471306-250MG
APOSOR471306-5G
D-tert-Leucine methyl ester hydrochloride
D-tert-Leucine methyl ester hydrochloride
Formel:
C₇H₁₅NO₂·ClH Molecular Weight: 181,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11113177 CAS-Nummer: 167223-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...