3-[N-(tert-Butyl)sulphamoyl]phenylboronic acid pinacol ester
Lieferant: Apollo Scientific
Synonyme:
3-[N-(tert-Butyl)sulphamoyl]phenylboronic Acid Pinacol Ester
OR471308-1G
OR471308-250MG
OR471308-5G
APOSOR471308-1GEA
397
EUR
APOSOR471308-1G
APOSOR471308-250MG
APOSOR471308-5G
3-[N-(tert-Butyl)sulphamoyl]phenylboronic acid pinacol ester
3-[N-(tert-Butyl)sulphamoyl]phenylboronic acid pinacol ester
Formel:
C₁₆H₂₆BNO₄S Molecular Weight: 339,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18384175 CAS-Nummer: 706820-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...