4-Hydroxyquinazoline-8-carbonitrile
Lieferant: Apollo Scientific
OR471314-1G
OR471314-250MG
APOSOR471314-1GEA
601
EUR
APOSOR471314-1G
APOSOR471314-250MG
4-Hydroxyquinazoline-8-carbonitrile
4-Hydroxyquinazoline-8-carbonitrile
Formel:
C₉H₅N₃O Molecular Weight: 171.156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646233 CAS-Nummer: 663194-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...