Ethyl Quinuclidine-4-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl quinuclidine-4-carboxylate
OR471319-1G
OR471319-250MG
APOSOR471319-1GEA
111
EUR
APOSOR471319-1G
APOSOR471319-250MG
Ethyl Quinuclidine-4-carboxylate
Ethyl Quinuclidine-4-carboxylate
Formel:
C₁₀H₁₇NO₂ Molecular Weight: 183.247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19686937 CAS-Nummer: 22766-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...