Methyl 4,4,4-Trichlorobutanoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 4,4,4-trichlorobutanoate
OR471320-1G
APOSOR471320-1GEA
81.2
EUR
APOSOR471320-1G
Methyl 4,4,4-Trichlorobutanoate
Methyl 4,4,4-Trichlorobutanoate
Formel:
C₅H₇Cl₃O₂ Molecular Weight: 205,467 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28144036 CAS-Nummer: 19376-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...