N-(2-Chloro-4-methyl-3-pyridyl)acetamide

Lieferant: Apollo Scientific

OR471339-1G OR471339-5G
APOSOR471339-1GEA 245 EUR
APOSOR471339-1G APOSOR471339-5G
N-(2-Chloro-4-methyl-3-pyridyl)acetamide
N-(2-Chloro-4-methyl-3-pyridyl)acetamide
Formel: C₈H₉ClN₂O
Molecular Weight: 184,623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21244018
CAS-Nummer: 76006-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR