N-Boc-2-phenylcyclopropylamine
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl (2-phenylcyclopropyl)carbamate
OR471350-1G
OR471350-250MG
APOSOR471350-1GEA
529
EUR
APOSOR471350-1G
APOSOR471350-250MG
N-Boc-2-phenylcyclopropylamine
N-Boc-2-phenylcyclopropylamine
Formel:
C₁₄H₁₉NO₂ Molecular Weight: 233,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21106315 CAS-Nummer: 92644-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...