(1R,2S)-2-Methylcyclohexylamine hydrochloride
Lieferant: Apollo Scientific
Synonyme:
(1R,2S)-2-Methylcyclohexylamine Hydrochloride
OR471352-1G
OR471352-250MG
APOSOR471352-1GEA
368
EUR
APOSOR471352-1G
APOSOR471352-250MG
(1R,2S)-2-Methylcyclohexylamine hydrochloride
(1R,2S)-2-Methylcyclohexylamine hydrochloride
Formel:
C₇H₁₅N·ClH Molecular Weight: 149,662 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201094 CAS-Nummer: 79389-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...