4-Hydroxy-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

OR471363-1G OR471363-250MG OR471363-5G
APOSOR471363-1GEA 713 EUR
APOSOR471363-1G APOSOR471363-250MG APOSOR471363-5G
4-Hydroxy-2-(hydroxymethyl)benzimidazole
4-Hydroxy-2-(hydroxymethyl)benzimidazole
Formel: C₈H₈N₂O₂
Molecular Weight: 164,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18807836
CAS-Nummer: 116345-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR