6-Chloro-2-(chloromethyl)quinazoline

Lieferant: Apollo Scientific

OR471380-1G OR471380-250MG OR471380-5G
APOSOR471380-1GEA 601 EUR
APOSOR471380-1G APOSOR471380-250MG APOSOR471380-5G
6-Chloro-2-(chloromethyl)quinazoline
6-Chloro-2-(chloromethyl)quinazoline
Formel: C₉H₆Cl₂N₂
Molecular Weight: 213.063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977344
CAS-Nummer: 1935563-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR