8-Bromo-2-chloro-5-methoxyquinoline-3-carbaldehyde

Lieferant: Apollo Scientific

OR471390-1G OR471390-5G OR471390-250MG
APOSOR471390-1GEA 429 EUR
APOSOR471390-1G APOSOR471390-5G APOSOR471390-250MG
8-Bromo-2-chloro-5-methoxyquinoline-3-carbaldehyde
8-Bromo-2-chloro-5-methoxyquinoline-3-carbaldehyde
Formel: C₁₁H₇BrClNO₂
Molecular Weight: 300,536 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18251403
CAS-Nummer: 1259319-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR