3-Hydroxy-3-methyl-2-(methylamino)butyric acid
Lieferant: Apollo Scientific
OR471435-1G
OR471435-250MG
APOSOR471435-1GEA
1010
EUR
APOSOR471435-1G
APOSOR471435-250MG
3-Hydroxy-3-methyl-2-(methylamino)butyric acid
3-Hydroxy-3-methyl-2-(methylamino)butyric acid
Formel:
C₆H₁₃NO₃ Molecular Weight: 147,172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18431958 CAS-Nummer: 108677-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...