1-Acetyl-3-(cyclopropylamino)piperidine

Lieferant: Apollo Scientific

OR471436-1G OR471436-250MG
APOSOR471436-1GEA 1940 EUR
APOSOR471436-1G APOSOR471436-250MG
1-Acetyl-3-(cyclopropylamino)piperidine
1-Acetyl-3-(cyclopropylamino)piperidine
Formel: C₁₀H₁₈N₂O
Molecular Weight: 182,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098078
CAS-Nummer: 1353986-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR