2-Methylbutyl Chloroformate

Lieferant: Apollo Scientific

OR471440-1G OR471440-25G OR471440-5G
APOSOR471440-1GEA 205 EUR
APOSOR471440-1G APOSOR471440-25G APOSOR471440-5G
2-Methylbutyl Chloroformate
2-Methylbutyl Chloroformate
Formel: C₆H₁₁ClO₂
Molecular Weight: 150,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28127255
CAS-Nummer: 20412-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR