(R)-3-(4-Bromophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific
Achtung

OR471446-5G OR471446-1G
APOSOR471446-5GEA 1010 EUR
APOSOR471446-5G APOSOR471446-1G
(R)-3-(4-Bromophenyl)-2-hydroxypropionic acid
(R)-3-(4-Bromophenyl)-2-hydroxypropionic acid
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977374
CAS-Nummer: 1141478-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR