4,7-Dihydroxyquinazoline
Lieferant: Apollo Scientific
OR471457-1G
OR471457-5G
APOSOR471457-1GEA
108
EUR
APOSOR471457-1G
APOSOR471457-5G
4,7-Dihydroxyquinazoline
4,7-Dihydroxyquinazoline
Formel:
C₈H₆N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01823180 CAS-Nummer: 16064-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...