3-(Methylamino)isonicotinic acid

Lieferant: Apollo Scientific

Synonyme: 3-(Methylamino)pyridine-4-carboxylic acid, 3-(Methylamino)isonicotinic acid

OR471460-1G OR471460-5G
APOSOR471460-1GEA 299 EUR
APOSOR471460-1G APOSOR471460-5G
3-(Methylamino)isonicotinic acid
3-(Methylamino)isonicotinic acid
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24160213
CAS-Nummer: 1461602-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR