Methyl 3-bromo-5-chloro-2-methylbenzoate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl 3-Bromo-5-chloro-2-methylbenzoate
OR471465-1G
OR471465-250MG
OR471465-5G
APOSOR471465-1GEA
549
EUR
APOSOR471465-1G
APOSOR471465-250MG
APOSOR471465-5G
Methyl 3-bromo-5-chloro-2-methylbenzoate
Methyl 3-bromo-5-chloro-2-methylbenzoate
Formel:
C₉H₈BrClO₂ Molecular Weight: 263.516 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24108598 CAS-Nummer: 1403597-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...