2-Chloro-7-hydroxyquinoline-3-carbaldehyde
Lieferant: Apollo Scientific
OR471468-1G
OR471468-250MG
APOSOR471468-1GEA
713
EUR
APOSOR471468-1G
APOSOR471468-250MG
2-Chloro-7-hydroxyquinoline-3-carbaldehyde
2-Chloro-7-hydroxyquinoline-3-carbaldehyde
Formel:
C₁₀H₆ClNO₂ Molecular Weight: 207.613 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03160579 CAS-Nummer: 519018-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...