6-Chloro-2,3-bis(hydroxymethyl)pyridine

Lieferant: Apollo Scientific
Achtung

OR471469-1G OR471469-250MG
APOSOR471469-1GEA 713 EUR
APOSOR471469-1G APOSOR471469-250MG
6-Chloro-2,3-bis(hydroxymethyl)pyridine
6-Chloro-2,3-bis(hydroxymethyl)pyridine
Formel: C₇H₈ClNO₂
Molecular Weight: 173,597 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18643364
CAS-Nummer: 220001-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR