6-Chloro-7-isopropyl-7H-purin-8(9H)-one

Lieferant: Apollo Scientific

OR471470-1G OR471470-250MG
APOSOR471470-1GEA 916 EUR
APOSOR471470-1G APOSOR471470-250MG
6-Chloro-7-isopropyl-7H-purin-8(9H)-one
6-Chloro-7-isopropyl-7H-purin-8(9H)-one
Formel: C₈H₉ClN₄O
Molecular Weight: 212,636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24615681
CAS-Nummer: 1226804-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR