5-Chloro-3-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
Lieferant: Apollo Scientific
OR471474-1G
OR471474-250MG
OR471474-5G
APOSOR471474-1GEA
601
EUR
APOSOR471474-1G
APOSOR471474-250MG
APOSOR471474-5G
5-Chloro-3-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
5-Chloro-3-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
Formel:
C₇H₆ClN₃O Molecular Weight: 183,595 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11559079 CAS-Nummer: 89660-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...