5-Chloro-1-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one

Lieferant: Apollo Scientific

OR471477-1G OR471477-250MG OR471477-5G
APOSOR471477-1GEA 997 EUR
APOSOR471477-1G APOSOR471477-250MG APOSOR471477-5G
5-Chloro-1-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
5-Chloro-1-methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one
Formel: C₇H₆ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD24563468
CAS-Nummer: 40852-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR