2-[2,4-Bis(benzyloxy)phenyl]acetic acid

Lieferant: Apollo Scientific

OR471482-1G OR471482-250MG
APOSOR471482-1GEA 601 EUR
APOSOR471482-1G APOSOR471482-250MG
2-[2,4-Bis(benzyloxy)phenyl]acetic acid
2-[2,4-Bis(benzyloxy)phenyl]acetic acid
Formel: C₂₂H₂₀O₄
Molecular Weight: 348.392 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664782
CAS-Nummer: 66056-40-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR