cis-3-(Boc-amino)-1-Cbz-4-(hydroxymethyl)pyrrolidine

Lieferant: Apollo Scientific

OR471486-1G OR471486-250MG OR471486-5G
APOSOR471486-1GEA 713 EUR
APOSOR471486-1G APOSOR471486-250MG APOSOR471486-5G
cis-3-(Boc-amino)-1-Cbz-4-(hydroxymethyl)pyrrolidine
cis-3-(Boc-amino)-1-Cbz-4-(hydroxymethyl)pyrrolidine
Formel: C₁₈H₂₆N₂O₅
Molecular Weight: 350.409 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28369193
CAS-Nummer: 246510-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR