2'-(1-Piperidinyl)acetophenone

Lieferant: Apollo Scientific

OR471487-1G OR471487-5G
APOSOR471487-1GEA 205 EUR
APOSOR471487-1G APOSOR471487-5G
2'-(1-Piperidinyl)acetophenone
2'-(1-Piperidinyl)acetophenone
Formel: C₁₃H₁₇NO
Molecular Weight: 203,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09755943
CAS-Nummer: 39911-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR