7-Bromo-4-methoxyindazole

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-4-methoxy-1H-indazole

OR471499-1G OR471499-250MG OR471499-5G
APOSOR471499-1GEA 804 EUR
APOSOR471499-1G APOSOR471499-250MG APOSOR471499-5G
7-Bromo-4-methoxyindazole
7-Bromo-4-methoxyindazole
Formel: C₈H₇BrN₂O
Molecular Weight: 227,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22380217
CAS-Nummer: 1337879-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR