(S)-3-Methyl-2-phenylbutanoic acid

Lieferant: Apollo Scientific

OR471506-1G OR471506-250MG OR471506-5G
APOSOR471506-1GEA 570 EUR
APOSOR471506-1G APOSOR471506-250MG APOSOR471506-5G
(S)-3-Methyl-2-phenylbutanoic acid
(S)-3-Methyl-2-phenylbutanoic acid
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00167435
CAS-Nummer: 13490-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR