8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane
Lieferant: Apollo Scientific
OR471512-1G
OR471512-5G
APOSOR471512-1GEA
154
EUR
APOSOR471512-1G
APOSOR471512-5G
8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane
8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane
Formel:
C₁₅H₁₇N₃O₄ Molecular Weight: 303,313 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18263009 CAS-Nummer: 28121-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...