2-(4-Bromo-1-pyrazolyl)ethyl Acetate
Lieferant: Apollo Scientific
OR471518-1G OR471518-5G
APOSOR471518-1GEA 306 EUR
APOSOR471518-1G APOSOR471518-5G
2-(4-Bromo-1-pyrazolyl)ethyl Acetate
2-(4-Bromo-1-pyrazolyl)ethyl Acetate
Formel: C₇H₉BrN₂O₂
Molecular Weight: 233.063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14707271
CAS-Nummer: 1215073-99-6