2-[2-(Benzyloxy)ethyl]oxirane

Lieferant: Apollo Scientific

OR471534-1G OR471534-5G
APOSOR471534-1GEA 244 EUR
APOSOR471534-1G APOSOR471534-5G
2-[2-(Benzyloxy)ethyl]oxirane
2-[2-(Benzyloxy)ethyl]oxirane
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27934088
CAS-Nummer: 94426-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR