5-Bromo-6-methylpyridine-2-carboxaldehyde
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-2-methyl-pyridine-6-carbaldehyde, 5-Bromo-6-methylpicolinaldehyde
OR471543-1G
OR471543-250MG
OR471543-5G
APOSOR471543-1GEA
499
EUR
APOSOR471543-1G
APOSOR471543-250MG
APOSOR471543-5G
5-Bromo-6-methylpyridine-2-carboxaldehyde
5-Bromo-6-methylpyridine-2-carboxaldehyde
Formel:
C₇H₆BrNO Molecular Weight: 200,033 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11227149 CAS-Nummer: 137778-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...