6-(Methylamino)pyridazine-3-carboxylic acid

Lieferant: Apollo Scientific

OR471544-1G OR471544-250MG
APOSOR471544-1GEA 1120 EUR
APOSOR471544-1G APOSOR471544-250MG
6-(Methylamino)pyridazine-3-carboxylic acid
6-(Methylamino)pyridazine-3-carboxylic acid
Formel: C₆H₇N₃O₂
Molecular Weight: 153,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656675
CAS-Nummer: 365413-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR