Methyl 7-methoxyindole-5-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR471552-1G
OR471552-250MG
APOSOR471552-1GEA
1060
EUR
APOSOR471552-1G
APOSOR471552-250MG
Methyl 7-methoxyindole-5-carboxylate 95%
Methyl 7-methoxyindole-5-carboxylate
Formel:
C₁₁H₁₁NO₃ Molecular Weight: 205,21 g/mol Schmelzpunkt: 176…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094980 CAS-Nummer: 180624-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...