6-Bromo-2,4-dichloro-8-methoxyquinazoline
Lieferant: Apollo Scientific
OR471555-1G
OR471555-250MG
APOSOR471555-1GEA
1420
EUR
APOSOR471555-1G
APOSOR471555-250MG
6-Bromo-2,4-dichloro-8-methoxyquinazoline
6-Bromo-2,4-dichloro-8-methoxyquinazoline
Formel:
C₉H₅BrCl₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD17392824 CAS-Nummer: 864292-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...