Methyl 3-bromo-5-chloro-4-methylbenzoate

Lieferant: Apollo Scientific

OR471567-1G OR471567-250MG OR471567-5G
APOSOR471567-1GEA 561 EUR
APOSOR471567-1G APOSOR471567-250MG APOSOR471567-5G
Methyl 3-bromo-5-chloro-4-methylbenzoate
Methyl 3-bromo-5-chloro-4-methylbenzoate
Formel: C₉H₈BrClO₂
Molecular Weight: 263.516 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11557543
CAS-Nummer: 203573-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR