Methyl 6-(Methylamino)pyridazine-3-carboxylate
Lieferant: Apollo Scientific
OR471568-1G
OR471568-250MG
OR471568-5G
APOSOR471568-1GEA
499
EUR
APOSOR471568-1G
APOSOR471568-250MG
APOSOR471568-5G
Methyl 6-(Methylamino)pyridazine-3-carboxylate
Methyl 6-(Methylamino)pyridazine-3-carboxylate
Formel:
C₇H₉N₃O₂ Molecular Weight: 167.165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12152730 CAS-Nummer: 1183150-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...