5-Chloro-2-iodoindole

Lieferant: Apollo Scientific

OR471570-250MG OR471570-1G OR471570-5G
APOSOR471570-250MGEA 248 EUR
APOSOR471570-250MG APOSOR471570-1G APOSOR471570-5G
5-Chloro-2-iodoindole
5-Chloro-2-iodoindole
Formel: C₈H₅ClIN
Molecular Weight: 277.489 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22563509
CAS-Nummer: 1388032-50-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR