1-Methylimidazole-5-boronic acid pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
OR471575-1G
OR471575-250MG
APOSOR471575-1GEA
102
EUR
APOSOR471575-1G
APOSOR471575-250MG
1-Methylimidazole-5-boronic acid pinacol ester
1-Methylimidazole-5-boronic acid pinacol ester
Formel:
C₁₀H₁₇BN₂O₂ Molecular Weight: 208.065 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11846349 CAS-Nummer: 942070-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...