2-Amino-4-methylnicotinic acid

Lieferant: Apollo Scientific
Achtung

OR471581-1G OR471581-250MG
APOSOR471581-1GEA 713 EUR
APOSOR471581-1G APOSOR471581-250MG
2-Amino-4-methylnicotinic acid
2-Amino-4-methylnicotinic acid
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427667
CAS-Nummer: 38076-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR