3-(Fmoc-amino)-3-(1-Boc-3-piperidyl)propanoic acid

Lieferant: Apollo Scientific

OR471583-1G OR471583-250MG OR471583-5G
APOSOR471583-1GEA 0 EUR
APOSOR471583-1G APOSOR471583-250MG APOSOR471583-5G
3-(Fmoc-amino)-3-(1-Boc-3-piperidyl)propanoic acid
3-(Fmoc-amino)-3-(1-Boc-3-piperidyl)propanoic acid
Formel: C₂₈H₃₄N₂O₆
Molecular Weight: 494,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425249
CAS-Nummer: 372144-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR