Methyl 1-(tert-butoxycarbonyl)-4-methoxyindoline-6-carboxylate
Lieferant: Apollo Scientific
OR471587-250MG
OR471587-1G
APOSOR471587-250MGEA
448
EUR
APOSOR471587-250MG
APOSOR471587-1G
Methyl 1-(tert-butoxycarbonyl)-4-methoxyindoline-6-carboxylate
Methyl 1-(tert-butoxycarbonyl)-4-methoxyindoline-6-carboxylate
Formel:
C₁₆H₂₁NO₅ Molecular Weight: 307,342 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30723198 CAS-Nummer: 928771-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...