Methyl 6-bromoindole-3-carboxylate 97%
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-bromo-1H-indole-3-carboxylate
OR471662-5G
APOSOR471662-5GEA
149
EUR
APOSOR471662-5G
Methyl 6-bromoindole-3-carboxylate 97%
Methyl 6-bromoindole-3-carboxylate
Formel:
C₁₀H₈BrNO₂ Molecular Weight: 254,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836005 CAS-Nummer: 868656-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...