3,4-Dihydrobenzo[b]azepine-2,5-dione 95%

Lieferant: Apollo Scientific

OR471699-1G OR471699-250MG
APOSOR471699-1GEA 111 EUR
APOSOR471699-1G APOSOR471699-250MG
3,4-Dihydrobenzo[b]azepine-2,5-dione 95%
3,4-Dihydrobenzo[b]azepine-2,5-dione
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02167972
CAS-Nummer: 16511-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR