1,2,3,4-Tetrahydroisoquinoline-6-carboxylic acid 95%

Lieferant: Apollo Scientific

OR471700-1G OR471700-250MG
APOSOR471700-1GEA 359 EUR
APOSOR471700-1G APOSOR471700-250MG
1,2,3,4-Tetrahydroisoquinoline-6-carboxylic acid 95%
1,2,3,4-Tetrahydroisoquinoline-6-carboxylic acid
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027511
CAS-Nummer: 933752-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR