4-Chlorobenzylboronic acid pinacol ester 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[(4-Chlorophenyl)methyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR471707-1G
APOSOR471707-1GEA 104 EUR
APOSOR471707-1G
4-Chlorobenzylboronic acid pinacol ester 95%
4-Chlorobenzylboronic acid pinacol ester
Formel: C₁₃H₁₈BClO₂
Molecular Weight: 252,545 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698531
CAS-Nummer: 475250-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR